Cole-Parmer TRS3X6 Tripod Bunsen Burner Stand, Cast Iron, 3 1/2" Ring, 6" Height
Representative image only.
Cole-Parmer TRS3X6 Tripod Bunsen Burner Stand, Cast Iron, 3 1/2" Ring, 6" Height
Cole-Parmer | Item: 3630057
Legs can be removed for easy storage
₹2,200.00INR / Each
₹2,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer TRS3X6   Tripod Bunsen Burner Stand, Cast Iron, 3 1/2" Ring, 6" Height
Cole-Parmer TRS3X6 Tripod Bunsen Burner Stand, Cast Iron, 3 1/2" Ring, 6" Height
3630057
₹2,200.00INR / Each
+ =
Total Price 6200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily position over burners Angled legs provide excellent stability
  • 1 year warranty
More About this Item
These single-ring tripod stands feature a cast-iron ring with black enamel finish and detachable zinc-plated legs. Ideal for supporting flat-bottom glassware over your bunsen burner.
1 Year Warranty
Specifications
  • Height (in)6
  • DescriptionTripod Bunsen Burner Stand, Cast Iron, 3 1/2" Ring