Cole-Parmer Titanium Tweezers Set of 5 w/ Hard Case
Representative image only.
Cole-Parmer Titanium Tweezers Set of 5 w/ Hard Case
Cole-Parmer | Item: 0738758
Ideal for handling microscopic particles
₹91,200.00INR / Set
₹91,200.00INR / Set
Product Description
Key Features
  • Chemical and corrosion resistant
  • Withstand temperatures up to 800°F (426°C)
  • Limited Lifetime Warranty
More About this Item
These lightweight, nonmagnetic titanium tweezers do not shed particles like stainless steel tweezers and are biocompatible. Typical applications include handling of microscopic particles, sensitive components in cleaning/chemical processes at high temperature, histology, and biology.
Limited Lifetime Warranty
Specifications
  • Overall Length (in)4 1/2
  • MaterialTitanium
  • SterileNo
  • DescriptionPrecision Titanium Tweezers Kit; 5 Piece Set
  • WarrantyLimited Lifetime