Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 3/4" Dia
Representative image only.
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 3/4" Dia
Cole-Parmer | Item: 0471041
For threaded probes only
₹11,400.00INR / Each
₹11,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 3/4" Dia
Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 3/4" Dia
0471041
₹11,400.00INR / Each
+ + =
Total Price 43200.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Autoclavable
  • 1 year warranty
More About this Item
The replaceable tip is for use with threaded probes and is fabricated from titanium alloy Ti-6Al-4V.
Use with threaded probe 04710-33.
1 Year Warranty
Specifications
  • Tip Diameter (in)0.75
  • Tip Diameter (cm)1.905
  • DescriptionThreaded Ultrasonic Probe Tip, Flat; 3/4" Dia