Cole-Parmer Threaded Ultrasonic Probe Extender for 1/2" Dia. Horn
Representative image only.
Cole-Parmer Threaded Ultrasonic Probe Extender for 1/2" Dia. Horn
Cole-Parmer | Item: 0471080
Use with tall or narrow vessels
₹69,700.00INR / Each
₹69,700.00INR / Each
Product Description
Key Features
  • Extenders are fabricated from titanium alloy TI-6AL-4V
  • Autoclavable
  • 1 year warranty
More About this Item
Extenders screw into threaded end probes of identical diameter in place of the replaceable tip. Recommended when working with tall narrow vessels such as Erlenmeyer flasks.
1 Year Warranty
Specifications
  • Tip Diameter (in)0.5
  • Tip Diameter (cm)1.27
  • DescriptionThreaded Ultrasonic Probe Extender for 1/2" Dia. Horn