Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, HDPE, 1/8" NPT(M) x 5/32" ID; 10/Pk
Representative image only.
Cole-Parmer Threaded to Hose Barb Fitting, Straight Adapter, HDPE, 1/8" NPT(M) x 5/32" ID; 10/Pk
Cole-Parmer | Item: 5974110
Threaded barb adapter design provides safe, difficult-to-break connection
₹1,300.00INR / Pkg of 10
₹1,300.00INR / Pkg of 10
Product Description
Key Features
  • deal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks.
These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialHDPE
  • Connection TypeNPT(M) x Hose Barb
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/8
  • Barb Size / Tubing Id (Port 2) (in)5/32
  • Flow PatternStraight
  • Regulatory ApprovalsRoHS;FDA
  • DescriptionThreaded to Hose Barb Fitting, Straight Adapter, HDPE, 1/8" NPT(M) x 5/32" ID; 10/Pk