Cole-Parmer Threaded to Hose Barb Fitting, 90° Elbow, Natural Nylon, 1/4" NPT(M) x 5/16" ID; 10/Pk
Representative image only.
Cole-Parmer Threaded to Hose Barb Fitting, 90° Elbow, Natural Nylon, 1/4" NPT(M) x 5/16" ID; 10/Pk
Cole-Parmer | Item: 5974182
Threaded barb adapter design provides safe, difficult-to-break connection
₹1,800.00INR / Pkg of 10
₹1,800.00INR / Pkg of 10
Product Description
Key Features
  • deal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
More About this Item
Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. The elbows feature a 90° bend and is designed to fit in a tight space. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialNylon
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Barb Size / Tubing Id (Port 2) (in)5/16
  • Flow Pattern90° Elbow
  • ColorNatural
  • Regulatory ApprovalsRoHS
  • DescriptionThreaded to Hose Barb Fitting, 90° Elbow, Natural Nylon, 1/4" NPT(M) x 5/16" ID; 10/Pk