Cole-Parmer Thermoplastic (SEBS) Tubing, 1/2" ID x 3/4" OD; 50 Ft

Representative image only.

Cole-Parmer Thermoplastic (SEBS) Tubing, 1/2" ID x 3/4" OD; 50 Ft

Cole-Parmer | Item: 5011981

Easily weldable/sealable—compatible with all common tube sealers and aseptic tube welders

₹53,600.00INR / Pkg of 1

₹53,600.00INR / Pkg of 1
Product Description

Key Features

  • Specifically designed for demanding biopharmaceutical applications
  • Does not contain BPA, phthalates, PVC, or peroxides
  • Sterilize by autoclave, gamma irradiation, and chemical sterilants
  • Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter <88> Class VI, Biological reactivity Tests, In Vivo (2020)
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • 1/2" ID x 3/4" OD
  • Manufacturer defect warranty

More About this Item

Designed for the most demanding biopharmaceutical applications, the thermoplastic SEBS (styrene-ethylene-butylene-styrene) tubing exhibits superior gas barrier properties and excellent pressure performance. Use the tubing in peristaltic pumps, single-use assemblies, process filling, and sampling devices.

Thermoplastic (SEBS) tubing does not contain BPA, phthalates, PVC, or peroxides that can leach out into the fluid, making it ideal for pharmaceutical applications, buffer and media preparation, and cell media processing. For an added level of particle control, a liner is placed between the tubing package and the box.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Tubing FormulationThermoplastic (SEBS)
  • Tubing OD (in)3/4
  • Tubing OD (mm)19.1
  • Tubing ID (in)1/2
  • Tubing ID (mm)12.7
  • Wall Thickness (in)1/8
  • Wall Thickness (mm)3.2
  • Clarity (translucent)Yes
  • Length (ft)50
  • Sterilization MethodGamma;Autoclave
  • Regulatory ApprovalsADCF;RoHS;Non-Phthalate;USP
  • Durometer68 Shore A
  • DescriptionThermoplastic (SEBS) Tubing, 1/2" ID x 3/4" OD; 50 Ft
Documents & Certificates

Certificate of Compliance

Reviews
Q&A