Cole-Parmer Temperature Compensating Probe for Ultrapure Water System, 1000 Ohm PT RTD
Representative image only.
Cole-Parmer Temperature Compensating Probe for Ultrapure Water System, 1000 Ohm PT RTD
Cole-Parmer | Item: 2700424
Optimize performance of your Ultrapure Water System
₹21,000.00INR / Each
₹21,000.00INR / Each
Product Description
Key Features
  • Easily measure the temperature of your ultrapure water
  • 1 year warranty
More About this Item
Temperature probe (upper left), pH electrode (upper right), and flow cell (bottom) shown in image.
Temperature probe (upper left), pH electrode (upper right), and flow cell (bottom) shown in image.
1 Year Warranty
Specifications
  • ApplicationsUltrapure
  • Temperature CompensationYes
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorSpade lugs
  • Cable Length (m)3
  • DescriptionTemperature Compensating Probe for Ultrapure Water System, 1000 Ohm PT RTD