Cole-Parmer TEMP INDICATORS -3-13C 1 EACH
Representative image only.
Cole-Parmer TEMP INDICATORS -3-13C 1 EACH
Cole-Parmer | Item: 9031605
Each style covers a wide range of temperatures, for use in many different application
₹320.00INR / Each
₹320.00INR / Each
Product Description
Key Features
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.