Cole-Parmer TELOS® SAX/PSA Multilayer SPE Column, 1g / 6mL, 30 / pk
Representative image only.
Cole-Parmer TELOS® SAX/PSA Multilayer SPE Column, 1g / 6mL, 30 / pk
Cole-Parmer | Item: 0647627
Achieve superior clean up when conducting multiresidue analysis
₹21,300.00INR / Pkg of 30
₹21,300.00INR / Pkg of 30
Product Description
Includes
Polyethylene frits
Key Features
  • Save money by extracting multiple suites with a single column
  • Clean up of extracts from fruits and vegetable samples prior to analysis of pesticides and herbicides
More About this Item
Multilayered SPE columns are designed for efficient extraction of multi-analyte suites. Layers of different SPE sorbents in a single SPE column allow a broader range of analytes to be extracted than is possible with a single sorbent. Each column features 20 µm frits at the top, bottom, and between sorbent layers. Columns feature Luer-slip outlet.
Columns are made of polypropylene.
Specifications
  • Bed Mass (mg)1000
  • End CapNo
  • Total Volume Capacity (mL)6
  • SorbentSAX/PSA