Cole-Parmer TELOS® QuEChERS Extraction Kit, AOAC 2007.01, 6g MgSO4 (Anhydrous) & 1.5g NaAc, 50 / pk
Representative image only.
Cole-Parmer TELOS® QuEChERS Extraction Kit, AOAC 2007.01, 6g MgSO4 (Anhydrous) & 1.5g NaAc, 50 / pk
Cole-Parmer | Item: 0647883
Quickly and economically extract the required analytes into solution
₹19,400.00INR / Pkg of 50
₹19,400.00INR / Pkg of 50
Buy It Together
Current Item
Cole-Parmer TELOS® QuEChERS Extraction Kit, AOAC 2007.01, 6g MgSO4 (Anhydrous) & 1.5g NaAc, 50 / pk
Cole-Parmer TELOS® QuEChERS Extraction Kit, AOAC 2007.01, 6g MgSO4 (Anhydrous) & 1.5g NaAc, 50 / pk
0647883
₹19,400.00INR / Pkg of 50
+
Cole-Parmer TELOS® QuEChERS Glass Homogenizer Beads, 50 / pk
0647857
₹9,700.00INR / Pkg of 50
=
Total Price 29100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Salt pack helps transfer salt to the centrifuge tube quickly and easily with minimal waste
  • Conform with documented AOAC 2007.01 and EN 15662 protocols
  • Break up complex sample types with homogenizer balls
More About this Item
TELOS QuEChERS extraction kits offer the first stage cleanup for many sample types. This initial crude extraction and partitioning step uses organic solvent and salt solution to extract the required analytes into solution. Select from extraction kits without a tube, kits with a 50 mL centrifuge tube, or kits with a tube and homogenizer beads. Beads aid in breaking up more complex sample types.
Specifications
  • Total Volume Capacity (mL)50
  • Salts6 g MgSO4 (anhydrous), 1.5 g NaAc.3H2O
  • MethodologyAOAC 2007.01
  • DescriptionKit without tubes