Cole-Parmer TELOS® PSA Media, 100g / bottle
Representative image only.
Cole-Parmer TELOS® PSA Media, 100g / bottle
Cole-Parmer | Item: 0647848
Save money by purchasing high quality sorbents in bulk quantities
₹36,600.00INR / Pkg of 1
₹36,600.00INR / Pkg of 1
Product Description
Key Features
  • Choose from a wide range of SPE media for packing your own columns
More About this Item
Pack your column with a single sorbent or create your own multilayer column using bulk media. Sorbents come in 100 g or 1 kg bottles.
Specifications
  • Media Mass (g)100
  • End CapNo
  • SorbentPSA (primary-secondary amine)