Cole-Parmer TELOS® Phase Separator, 15 mL; 100/pk
Representative image only.
Cole-Parmer TELOS® Phase Separator, 15 mL; 100/pk
Cole-Parmer | Item: 0647549
Quickly and easily separate aqueous and chlorinated solvents
₹18,000.00INR / Pkg of 100
₹18,000.00INR / Pkg of 100
Product Description
Key Features
  • Gravity flow filtration process, no vacuum required
  • No need for separating funnels and rewashing glassware
More About this Item
Simply apply aqueous and chlorinated solvents to the column for a quick separation. A special hydrophobic frit allows the chlorinated solvent to pass through under gravity while preventing the aqueous layer from flowing though. The aqueous layer remains in the separator column, separating the two phases and allowing additional work up.
Specifications
  • Total Volume Capacity (mL)15