Cole-Parmer TELOS® PCX MicroPlate™ 96-well Plate, loose wells, 5mg / 1mL, 100 / pk
Representative image only.
Cole-Parmer TELOS® PCX MicroPlate™ 96-well Plate, loose wells, 5mg / 1mL, 100 / pk
Cole-Parmer | Item: 0647414
Populate base plate with only the wells needed
₹44,900.00INR / Pkg of 100
₹44,900.00INR / Pkg of 100
Buy It Together
Current Item
Cole-Parmer TELOS®  PCX MicroPlate™ 96-well Plate, loose wells, 5mg / 1mL, 100 / pk
Cole-Parmer TELOS® PCX MicroPlate™ 96-well Plate, loose wells, 5mg / 1mL, 100 / pk
0647414
₹44,900.00INR / Pkg of 100
+
Cole-Parmer TELOS® MicroPlate Sealing Strips, 24 / pk
0647411
₹5,700.00INR / Pkg of 24
=
Total Price 50600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 50 µL elution volume for minimal sample dilution and reduced evaporation time
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
  • Applications: Mixed-mode strong cation exchange sorbent for basic analyte extraction
More About this Item
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® neo™ MicroPlate™ SPE loose wells allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µL.
Use the loose wells for assays when the sample numbers vary, populating the base plate with only the required number of wells. Base plates are required to use loose wells. Full or partially populated plates can be processed using either vacuum or positive pressure. Order method development kit to build a method development plate to suit your application.
Use the loose-well plates for assays when the sample numbers vary, populating the base plate with only the required number of wells. Base plates are required to use loose-well plates. Order method development kit to build a method development plate to suit your applicatiion.
Specifications
  • Bed Mass (mg)5
  • Column Particle Size (µm)60
  • Bottom TypePolymer based
  • ApplicationsMixed-mode strong cation exchange
  • SorbentPCX
  • DescriptionPCX MicroPlate SPE Microplate, loose wells, 5 mg sorbent; 100/pk