Cole-Parmer TELOS® Neutral Matrix SLE 96-well Plate, pH 7, 200mg / 2mL, 1 / pk
Representative image only.
Cole-Parmer TELOS® Neutral Matrix SLE 96-well Plate, pH 7, 200mg / 2mL, 1 / pk
Cole-Parmer | Item: 0648233
Perfectly suited for automation and high throughput procedures
₹60,700.00INR / Each
₹60,700.00INR / Each
Product Description
Key Features
  • Efficient extraction
  • High analyte recovery with no emulsion formation
  • Fixed-well plate
  • pH 7
  • 200 mg
More About this Item
Use supported liquid extraction plates when you need to extract samples quickly and effectively. Wells are packed with a diatomaceous earth, a neutral matrix, to provide support to absorb the aqueous sample. Once the aqueous sample is fully absorbed, a water immiscible organic solvent is used to elute the analytes. High analyte recoveries are obtained without any offline steps such as protein precipitation, and sample extracts are free of proteins and phospholipids. A hydrophilic top frit allows the sample to pass through while minimizing any blockage.
Use neutral matrix columns (NM) in analytical applications such as isolating drugs from biological fluids.
Specifications
  • Bed Mass (mg)200
  • ApplicationsNeutral matrix
  • DescriptionSLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 7, 200 mg; 1/EA