Cole-Parmer TELOS® Multilayer Sodium Sulfate/Florisil SPE Column, 2g / 6mL, 30 / pk
Representative image only.
Cole-Parmer TELOS® Multilayer Sodium Sulfate/Florisil SPE Column, 2g / 6mL, 30 / pk
Cole-Parmer | Item: 0647639
Achieve superior clean up when conducting multiresidue analysis
₹28,200.00INR / Pkg of 30
₹28,200.00INR / Pkg of 30
Product Description
Includes
Polyethylene frits
Key Features
  • Save money by extracting multiple suites with a single column
  • Remove water from water immiscible solvents
More About this Item
Multilayered SPE columns are designed for efficient extraction of multi-analyte suites. Layers of different SPE sorbents in a single SPE column allow a broader range of analytes to be extracted than is possible with a single sorbent. Each column features 20 µm frits at the top, bottom, and between sorbent layers. Columns feature Luer-slip outlet.
Columns are made of polypropylene.
Specifications
  • Bed Mass (mg)2000
  • End CapNo
  • Total Volume Capacity (mL)6
  • SorbentNa₂SO₄/MgO₃Si