Cole-Parmer TELOS® Luer Cap
Representative image only.
Cole-Parmer TELOS® Luer Cap
Cole-Parmer | Item: 0647975
Optimize performance of your TELOS chromatography columns
₹3,700.00INR / Pkg of 100
₹3,700.00INR / Pkg of 100
Product Description
Key Features
  • Choose from a wide range of accessories for flash chromatography dry loading, filtration, and large volume processing
  • Easily cap columns with Luer connections
More About this Item
Provide a secure seal for both the inlet and outlet of TELOS flash columns with the luer cap and plug. TELOS column luer cap can be used in conjunction with the appropriate size column for either room temperature reactions or short-term compound storage.
Easily and quickly adapt 1-, 3-, or 6-mL TELOS SPE columns the column adapter.
Specifications
  • ColorBlack
  • Compatible WithFlash chromatography columns
  • ConnectionsLuer
  • DescriptionTELOS Luer Cap