Cole-Parmer TELOS® Endcapped Multilayer SPE Column, C18 / ENV, 700 mg sorbent, 6 mL; 30/pk
Representative image only.
Cole-Parmer TELOS® Endcapped Multilayer SPE Column, C18 / ENV, 700 mg sorbent, 6 mL; 30/pk
Cole-Parmer | Item: 0647809
Achieve superior clean up when conducting multiresidue analysis
₹32,900.00INR / Pkg of 30
₹32,900.00INR / Pkg of 30
Product Description
Includes
Polyethylene frits
Key Features
  • Save money by extracting multiple suites with a single column
  • Extract a wide range of pesticides and herbicides from water
More About this Item
Multilayered SPE columns are designed for efficient extraction of multi-analyte suites. Layers of different SPE sorbents in a single SPE column allow a broader range of analytes to be extracted than is possible with a single sorbent. Each column features 20 µm frits at the top, bottom, and between sorbent layers. Columns feature Luer-slip outlet.
Columns are made of polypropylene.
Specifications
  • Bed Mass (mg)700
  • End CapYes
  • Total Volume Capacity (mL)6
  • SorbentC18/ENV (endcapped)