Cole-Parmer TELOS® DNPH coated silica SPE Cartridge, 350 mg / 1 mL, 50 / pk
Representative image only.
Cole-Parmer TELOS® DNPH coated silica SPE Cartridge, 350 mg / 1 mL, 50 / pk
Cole-Parmer | Item: 0647632
Extract polar analytes from nonaqueous matrices
₹177,000.00INR / Pkg of 50
₹177,000.00INR / Pkg of 50
Product Description
Includes
20 µm top and bottom polyethylene frits
Key Features
  • Constructed of durable polypropylene
More About this Item
Use the TELOS polar (normal phase) SPE cartridges to retain analytes of interest or to remove polar interferences from the sample. Columns with Florisil® PR grade (magnesium silicate) sorbent have larger particle size than NH2 and are commonly used for lipid separation and pesticide extraction. All columns are made of polypropylene and feature female Luer-slip inlet and male Luer-slip outlet.