Cole-Parmer TELOS® Acidic Alumina (AL-A) SPE Column, 50mg / 1mL, 100 / pk
Representative image only.
Cole-Parmer TELOS® Acidic Alumina (AL-A) SPE Column, 50mg / 1mL, 100 / pk
Cole-Parmer | Item: 0647641
Easily isolate compounds from nonpolar solvents
₹26,000.00INR / Pkg of 100
₹26,000.00INR / Pkg of 100
Product Description
Includes
20 µm polyethylene top and bottom frits.
Key Features
  • Polar SPE sorbents
More About this Item
Alumina (acidic, basic, and neutral) are polar SPE sorbents commonly used to isolate compounds from nonpolar solvents. Columns are made of polypropylene and feature Luer-slip outlet.
Specifications
  • Bed Mass (mg)50
  • Total Volume Capacity (mL)1
  • SorbentAlumina—acidic (AL-A)