Cole-Parmer TELOS® 20µm Polyethylene Top Frit, 1/8" for 4g Flash Column, 100 / pk

Representative image only.

Cole-Parmer TELOS® 20µm Polyethylene Top Frit, 1/8" for 4g Flash Column, 100 / pk

Cole-Parmer | Item: 0647532

Extend the life of your dry loading cartridges with these replacement parts

₹21,500.00INR / Pkg of 100

₹21,500.00INR / Pkg of 100
Product Description

Key Features

  • Constructed of durable polyethylene
  • Compatible with TELOS 4 g dry loading cartridge with slip tip
  • 20 µ fritted disk

More About this Item

Choose from a variety of replacement top frit sizes for your Cole-Parmer TELOS dry loading columns.

Specifications
  • MaterialPolyethylene
  • Compatible WithColumn 06475-53; 20 µm
  • DescriptionTop Frit for Dry Loading Cartridge, Slip Tip, 4 g, 20 µm; 100/pk
Reviews
Q&A