Cole-Parmer TC DISH PETRI 35MM NT-S 960/CS
Representative image only.
Cole-Parmer TC DISH PETRI 35MM NT-S 960/CS
Cole-Parmer | Item: 1597132
Excellent optical quality helps to promote healthy cells and reproducible results
₹23,700.00INR / Case of 1
₹23,700.00INR / Case of 1
Product Description
Key Features
  • Select from non-treated and surface-treated dishes with 7 different diameters of 1.8, 3.5, 6.0, 7.0, 9.0, 10.0, and 15.0 cm
  • Flat bottom with uniform wall thickness ensures stability
  • Corrugation free and smooth dish surface maximizes usable growth area
  • The rim on upper side of the lid mates with the dish brim for easy and secure stacking
  • Lids with vents are available for very effective gas exchange
  • Gamma sterilized and non-pyrogenic
  • Packaged in durable zip re-sealable self-standing plastic bags that allow dishes to remain upright and reduce contamination
  • Inner packaging is individually labelled for lot traceability
Specifications
  • Surface TreatmentGeneral, non-treated
  • Lid (Included)Yes
  • Membrane Growth Area (cm²)8.5
  • Description10 dishes/bag; 96 bags/case Total: 960 dishes