Cole-Parmer TC 12well NT-S100/cs
Representative image only.
Cole-Parmer TC 12well NT-S100/cs
Cole-Parmer | Item: 1597261
Extremely smooth surface and raised well rims reduce evaporation and maximize results
₹17,100.00INR / Case of 1
₹17,100.00INR / Case of 1
Buy It Together
Current Item
Cole-Parmer TC 12well NT-S100/cs
Cole-Parmer TC 12well NT-S100/cs
1597261
₹17,100.00INR / Case of 1
+ =
Total Price 74300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Gamma sterilized and nonpyrogenic
  • Individually packaged in peel-to-open blister pack
  • Printed lot numbers on both plate and packaging for effortless traceability
  • Optically clear plates for easy visualization of cell growth
More About this Item
Use these sterile well plates to serve your various cell and tissue culture needs. With individual condensation rings and a one-way, tight-fitting lid, they reduce cross-contamination and help prevent evaporation. An alphanumeric notation on each well aids in simple identification. Transparent polystyrene construction allows for easy visualization of cell growth and facilitates observation under a microscope.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment for optimal quality, these cell and tissue culture well plates are individually packaged in peel-to-open blister packs, sterilized by irradiation (SAL 10-6), and are DNase/RNase free, nonpyrogenic, and noncytotoxic. They conform to USP Class VI and are quality traceable with the lot number on both the plate and packaging.
Plates with a treated surface are suitable for common adherent cell lines due to the excellent hydrophilicity of the surface while untreated plates are suitable for suspension cell culture. Compatible with most multi-well plate instruments and equipment.
Sterile
Specifications
  • Number of Wells12
  • Max Working Well Volume (mL)6.8
  • Lid (Included)Yes
  • SterileYes
  • DescriptionGeneral, non-treated