Cole-Parmer T30-2-506 Thermocouple, Wire, Type-T, 30-Gauge, FEP Insulation, 100-ft Spool
Representative image only.
Cole-Parmer T30-2-506 Thermocouple, Wire, Type-T, 30-Gauge, FEP Insulation, 100-ft Spool
Cole-Parmer | Item: 0854104
Make your own extension cables
₹20,400.00INR / Each
₹20,400.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire comes in 30 gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with fluorinated ethylene propylene resin (FEP/FEP) or fiberglass materials as well as in 100-ft or 1000-ft lengths for any application.
6 Month Warranty
Specifications
  • Connector TypeT
  • MaterialFluorinated ethylene propylene (FEP)
  • Wire Gauge (Ga)30
  • Error Limit±1.5% reading from -330 to -85°F (-200 to -65°C); ±1.8°F (±1°C) from -85 to 270°F (-65 to 130°C); ±0.75% reading from 270 to 660°F (130 to 350°C)
  • Inner InsulationFluorinated ethylene propylene (FEP)
  • Outer InsulationFluorinated ethylene propylene (FEP)
  • Stranded / SolidSolid
  • Special InformationSPECIAL LIMITS OF ERROR
  • Min Temperature (° F)-328
  • Max Temperature (° F)400
  • Min Temperature (° C)-200
  • Max Temperature (° C)204
  • Length (ft)100
  • Size (in)0.030 in x 0.048 in
  • DescriptionThermocouple, Wire, Type-T, 30-Gauge, FEP Insulation, 100-ft Spool