Cole-Parmer Swivel Clamp, Two-Prong, Medium, Nickel-Plated Zinc
Representative image only.
Cole-Parmer Swivel Clamp, Two-Prong, Medium, Nickel-Plated Zinc
Cole-Parmer | Item: 0804702
Provide support for your specific laboratory equipment
₹7,000.00INR / Each
₹7,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Ideal for cylindrical shaped labware. Swivel action allows full 360° adjustment. Clamps include integral connector and both slip-on vinyl and fiberglass sleeves.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc with red vinyl and black fiberglass sleeves
  • Clamp TypeEquipment, Two- or Three-Prong
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)3
  • Length (in)6.06
  • DescriptionSwivel Clamp, Two-Prong, Medium, Nickel-Plated Zinc