Cole-Parmer Swivel Clamp, Three-Prong, Medium, Stainless Steel
Representative image only.
Cole-Parmer Swivel Clamp, Three-Prong, Medium, Stainless Steel
Cole-Parmer | Item: 0812991
Provide support for your specific laboratory equipment
₹52,500.00INR / Each
₹52,500.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Ideal for cylindrical shaped labware. Swivel action allows full 360° adjustment. Clamps include integral connector and both slip-on vinyl and fiberglass sleeves.
Two year warranty
Specifications
  • Clamp MaterialStainless Steel
  • Clamp TypeEquipment, Swivel
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)2.72
  • Accepts Rod Diameter (in)0.5
  • Length (in)6.06
  • DescriptionSwivel Clamp, Three-Prong, Medium, Stainless Steel