Cole-Parmer Suspension Clamp, Nickel-Plated Zinc, 5.39" L

Representative image only.

Cole-Parmer Suspension Clamp, Nickel-Plated Zinc, 5.39" L

Cole-Parmer | Item: 0802708

Provide support for your specific laboratory equipment

₹6,200.00INR / Each

₹6,200.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Ideal for suspending thermometers, potash bulbs or drying tubes. Integrated connector included.

1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeEquipment, Fixed Suspension
  • Length (in)5.39
  • DescriptionSuspension Clamp, Nickel-Plated Zinc, 5.39" L
  • Warranty1 year
Reviews
Q&A