Cole-Parmer Strip Tube Rotor, 2 x 0.2-mL 8-Tube Strips
Representative image only.
Cole-Parmer Strip Tube Rotor, 2 x 0.2-mL 8-Tube Strips
Cole-Parmer | Item: 8523006
Enhance the performance of your Cole-Parmer® CF-200 Microcentrifuge
₹93,600.00INR / Each
₹93,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer Strip Tube Rotor, 2 x 0.2-mL 8-Tube Strips
Cole-Parmer Strip Tube Rotor, 2 x 0.2-mL 8-Tube Strips
8523006
₹93,600.00INR / Each
+ =
Total Price 333600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 3 year warranty
  • Previous part number SCF3/2
More About this Item
The Cole-Parmer CF-200 microcentrifuge includes a fixed-angle rotor that accommodates up to twelve 1.5- or 2.0-mL tubes. Order tube adapters to used 0.2- or 0.5-mL tubes, or order the strip tube rotor to accommodate two 8-place 0.2-mL strip tubes.
3 Year Warranty
Specifications
  • Rotor TypeFixed Angle
  • Tube Sizes Accepted0.2 mL
  • Number Of Tubes Accepted16
  • Adapter Shape TypeConical
  • DescriptionStrip Tube Rotor, 2 x 0.2-mL 8-Tube Strips