Cole-Parmer Standard Support Holder, Stainless Steel; with 2 openings
Representative image only.
Cole-Parmer Standard Support Holder, Stainless Steel; with 2 openings
Cole-Parmer | Item: 0812982
Designed with over sized, easy to grip knobs for precise tightening
₹29,900.00INR / Each
₹29,900.00INR / Each
Product Description
Key Features
  • Secure your motor to a support stand with easy
  • Constructed of durable stainless steel
  • 2 year warranty
More About this Item
Accommodates 1/2" (12.7mm) diameter shaft of a motor mounting rod and 5/8" (16mm) diameter support stand rod. Motor and support stand sold separately.
Two year warranty
Specifications
  • Clamp MaterialStainless Steel
  • Clamp TypeFrame and Rod
  • DescriptionRegular Holder, Stainless Steel (Standard Support holder with 2 openings)