Cole-Parmer Stainless Steel Wire Shelf, for 5.3/5.4 cu ft units
Representative image only.
Cole-Parmer Stainless Steel Wire Shelf, for 5.3/5.4 cu ft units
Cole-Parmer | Item: 7951020
Add additional shelf space with these stainless steel wire racks
₹39,500.00INR / Each
₹39,500.00INR / Each
Product Description
Key Features
  • Use with 5.4 cu ft Digitally Controlled Mechanical Convection Incubators #79510-16 and -18
  • Use with 5.3 cu ft Economy Mechanical Convection Ovens #52402-91 and -94
  • Use with 5.3 cu ft Mechanical Convection Ovens #52402-85 and -88
  • Use with 5.3 cu ft Mechanical Convection Ovens #52412-78 and -81
  • Use with 5.3 cu ft Digital Gravity Convection Ovens #52412-85 and -88
  • Use with 5.3 cu ft Economy Mechanical Convection Ovens #52412-91 and -94
  • 2 year warranty
More About this Item
Electropolished stainless steel shelves are easy to remove without tools and easy to clean.
Two year warranty
Specifications
  • MaterialStainless steel
  • Length (InchShort)6 1/2
  • Width (InchShort)22 23/32
  • DescriptionStainless Steel Wire Shelf, for 5.3/5.4 cu ft units
  • Warranty2 Years