Cole-Parmer Solid Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
Representative image only.
Cole-Parmer Solid Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
Cole-Parmer | Item: 0885041
Achieve optimal cleaning results
₹36,200.00INR / Each
₹36,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Solid Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
Cole-Parmer Solid Stainless Steel Insert Tray for 0.5 Gal Ultrasonic Cleaners
0885041
₹36,200.00INR / Each
+
Elmasonic Ultrasonic Cleaner Basket, E+ Series; 0.25 gal.
5998835
₹16,400.00INR / Each
=
Total Price 52600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Supports items that require indirect cleaning
  • Prevents damage to transducers
  • 1 year warranty
More About this Item
Made of 304 stainless steel, these solid insert trays can be used to support items that require indirect cleaning. Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers.
Never use ultrasonic cleaners with flammable solvents. A cleaning solution must be used with these cleaners. Use an insert tray or beaker positioning cover and beaker with these ultrasonic cleaners. Ultrasonic acticity is greatest starting about an inch from the bottom of the tank.
1 Year Warranty
Specifications
  • Capacity (Gal)0.5
  • Capacity (Liters)1.9
  • Length (InchShort)4 1/2
  • Width (InchShort)4 1/2
  • Height (InchShort)3
  • Length (cm)11.43
  • Width (cm)11.43
  • Height (cm)7.62
  • Material304SS
  • DescriptionSolid tray for 1/2 Gallon and Stainless Steel 3/4 Gallon Cleaners.