Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 8; 10/Pk
Representative image only.
Cole-Parmer Solid Green Neoprene Stoppers, Standard Size 8; 10/Pk
Cole-Parmer | Item: 6299126
₹13,500.00INR / Pkg of 10
₹13,500.00INR / Pkg of 10
Product Description
More About this Item
Green neoprene stoppers withstand petroleum products, fats, oils, and organic chemicals. Stoppers exhibit good resiliency and shelf-life. Each stopper has its size in raised figures on top. Contain no sulfur.Autoclavable.
Specifications
  • Standard Size Number8
  • Top Outside Diameter (OD) (mm)41
  • Bottom Outside Diameter (OD) (mm)33
  • Height (mm)25
  • ColorGreen
  • MaterialNeoprene
  • DescriptionSolid Green Neoprene Stoppers, Standard Size 8; 10/Pk