Cole-Parmer Silver Electrodes, Anode; 3/Pk
Representative image only.
Cole-Parmer Silver Electrodes, Anode; 3/Pk
Cole-Parmer | Item: 0265634
Ensure accurate results with replacement anode, cathode, and detecting electrodes for chlorine analyzer
₹45,900.00INR / Pkg of 3
₹45,900.00INR / Pkg of 3
Product Description
Key Features
  • Discard when the length is less than 57 mm
  • Maintain electrodes with cleaning polish 02656-40
More About this Item
Do not leave the electrodes immersed in reagents when the instrument is not in use.
Specifications
  • Includes3 anodes
  • DescriptionSilver Electrodes, Anode. Pack Of Three