Cole-Parmer Shelf Grip Clamp; Nickel-Plated Zinc
Representative image only.
Cole-Parmer Shelf Grip Clamp; Nickel-Plated Zinc
Cole-Parmer | Item: 0805000
Provide support for your specific laboratory equipment
₹52,800.00INR / Each
₹52,800.00INR / Each
Product Description
Key Features
  • 3 month warranty
More About this Item
Designed to affix to a bench or shelf up to 2” (51mm) thick, and attach via the integrated connector a standard 1⁄2” (1.3cm) clamp support rod. The maximum load for this clamp is 150lbs (68kg).
2" max shelf, holds 2 rods
3 Month Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeFrame and Rod
  • Max Grip Range (InchShort)2
  • Accepts Rod Diameter (in)0.5
  • Length (in)7.75
  • DescriptionShelf Grip Clamp; Nickel-Plated Zinc