Cole-Parmer Serological Pipette and Organizing Rack Bundle, Polystyrene Self Aspirating Pipettes, 2 mL, Sterile, Individually Wrapped; 2 x 500/CS
Representative image only.
Cole-Parmer Serological Pipette and Organizing Rack Bundle, Polystyrene Self Aspirating Pipettes, 2 mL, Sterile, Individually Wrapped; 2 x 500/CS
Cole-Parmer | Item: 2530708
Protect pipettes and minimize clutter
₹79,200.00INR / Each
₹79,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Serological Pipette and Organizing Rack Bundle, Polystyrene Self Aspirating Pipettes, 2 mL, Sterile, Individually Wrapped; 2 x 500/CS
Cole-Parmer Serological Pipette and Organizing Rack Bundle, Polystyrene Self Aspirating Pipettes, 2 mL, Sterile, Individually Wrapped; 2 x 500/CS
2530708
₹79,200.00INR / Each
+ +
BrandTech BLAUBRAND®  Bulb Pipette, Glass, Class A, 20 mL; 6/PK
2521029
₹32,100.00INR / Pkg of 6
=
Total Price 130300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
1000 sterile serological pipettes and one pipette organizing rack.
Key Features
  • Bundle combines serological pipettes and organizing rack for added convenience
  • Pipettes are sterilized by gamma irradiation and individually wrapped to eliminate cross contamination
  • Safely store and protect your pipettes with this durable acrylic rack
  • Pipettes are tested for endotoxins, cytotoxicity, and are guaranteed nonhemolytic
More About this Item
These Cole-Parmer® serological pipettes and organizing rack are bundled for exceptional convenience. Heavy-duty, blue-tinted, acrylic rack features five angled compartments which can house 1 to 100 mL glass or plastic pipettes. Two slots at the top of the rack are designed to hold pipette controllers. Organizing rack has exterior dimensions of 3.5” W x 15.875” H x 11.5” D.
These serological pipettes are made from transparent polystyrene (PS) and sterilized by gamma irradiation. Choose from standard serological pipettes with highly legible permanent graduations or aspirating serological pipettes without graduations and filter plugs for vacuum aspirating and free-flowing applications.
Plastic serological pipettes are supplied without graduations and filter plugs for vacuum aspirating and free-flowing applications. They are ideal for tissue culture applications when aspirating larger volumes from dishes, flasks, or roller bottles. Individually wrapped.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • SterileYes
  • Max Volume (mL)2
  • Graduation IntervalNone
  • Overall Length (InchShort)12
  • Tip StyleSelf Aspirating
  • Body MaterialAcrylic Rack
  • Wetted MatterialPolystyrene Pipettes
  • Color CodeTransparent
  • Filter TypeNone
  • Package Of1000 Individually wrapped pipettes
  • DescriptionSerological Pipette and Organizing Rack Bundle, Polystyrene Self Aspirating Pipettes, 2 mL, Sterile, Individually Wrapped; 2 x 500/CS