Cole-Parmer Semi-Micro pH Electrode, Glass/3 ft Cable; BNC
Representative image only.
Cole-Parmer Semi-Micro pH Electrode, Glass/3 ft Cable; BNC
Cole-Parmer | Item: 1594555
Easily measure samples as small as 200uL with great precision
₹29,700.00INR / Each
₹29,700.00INR / Each
Product Description
Includes
3.3-ft (1-m) cable and BNC connector.
Key Features
  • Unique sensor technology for faster response and increased stability
  • Fits into small vessels, sensing tip diameter is 6 mm for minimum sample volume of 200 uL
  • Response time is 0.01 pH units in 10 seconds
  • Compatible with most brands of pH meters with standard BNC connection
  • Easily measure samples as small as 200uL with great precision
  • Unique sensor technology for faster response and increased stability
  • Fits into small vessels, sensing tip diameter is 6 mm for minimum sample volume of 200uL
  • Response time is 0.01 pH units in 10 seconds
  • Compatible with most brands of pH meters with standard BNC connection
More About this Item
This refillable semi-microelectrode features fast, accurate, and stable pH readings. Readings stabilize to 0.01 pH units in 10 seconds. Electrodes measure pH from 0 to 14. Maximum temperature is 70°C. Tip is designed to fit into microtubes and other liquid containers commonly used in the laboratory.
Specifications
  • DescriptionSemi-Micro pH Electrode, Glass/3 ft Cable; BNC