Cole-Parmer SDS-7015 Standard cell refill solution, 125 mL
Representative image only.
Cole-Parmer SDS-7015 Standard cell refill solution, 125 mL
Cole-Parmer | Item: 2740074
Accessories for the Cole-Parmer pH and ORP differential sensors
₹9,500.00INR / Each
₹9,500.00INR / Each
Product Description
Key Features
  • Quick and easy maintenance with optional replacement salt bridge
More About this Item
Sensor life is extended with the replacement salt bridge feature and refill solution, which avoids having to replace the entire sensor. Choose from the replacement salt bridge in packs of either 1, 3, 10 10 each. The standard cell refill solution is available in three sizes as well.
1 Year Warranty
Specifications
  • Accessory TypeStandard cell refill solution
  • MaterialPPS
  • DescriptionStandard cell refill solution, 125 mL