Cole-Parmer Screw Cap, Polypropylene, White, 24 mm, Closed, No Septa;1000/pk
Representative image only.
Cole-Parmer Screw Cap, Polypropylene, White, 24 mm, Closed, No Septa;1000/pk
Cole-Parmer | Item: 9870387
No more damaged autosampler needles or sample loss
₹20,100.00INR / Pkg of 1000
₹20,100.00INR / Pkg of 1000
Customers Also Bought
Product Description
Key Features
  • Ensure septa is not deformed due to overtightening and extend the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond­® septum to avoid sample contamination
More About this Item
Select from polypropylene (PP), polyethylene (PE) or magnetic silver caps with various septum materials. Caps are available in different sizes, colors, septum materials, and open or closed configurations to suit your application. UltraBond cap and septum form a one-piece unit with a bonding process without adhesives for contamination-free results. Pre-slit septa prevent a vacuum forming inside the vial.
This closed polypropylene cap does not include a septa. You can choose from a wide range of septas for your application or our EPA quality septa 98700-75.
Please Note: Vials, caps, and septa formerly under the Kinesis brand have now been rebranded Cole-Parmer. All the same great features and specifications, just with a new and improved look.
Specifications
  • Cap TypeScrew Cap
  • Cap Diameter (mm)24
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Septum TypeNone
  • SterileNo
  • Special FeaturesClosed Cap No Septa
  • DescriptionScrew Cap, Polypropylene, White, 24 mm, Closed, No Septa;1000/pk