Cole-Parmer SBL-2 Basket for Fluidized Sand Bath SBL-2
Representative image only.
Cole-Parmer SBL-2 Basket for Fluidized Sand Bath SBL-2
Cole-Parmer | Item: 0118485
₹69,500.00INR / Each
₹69,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer SBL-2 Basket for Fluidized Sand Bath SBL-2
Cole-Parmer SBL-2 Basket for Fluidized Sand Bath SBL-2
0118485
₹69,500.00INR / Each
+ + + =
Total Price 748500.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Durable stainless steel mesh construction
  • Safely hold samples away from heating elements
  • Easily retrieve samples from the bath
  • 1 year warranty
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.
Stainless steel baskets are available for each SB model bath to prenvent workpieces from touching the heater elements, allow good flow of Alundum through the basket and to make retrieval of parts easier.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithSBL-2 Fluidized sand baths
  • DescriptionBasket for Fluidized Sand Bath SBL-2