Cole-Parmer Sanitary End Cap, Polypropylene, 3"
Representative image only.
Cole-Parmer Sanitary End Cap, Polypropylene, 3"
Cole-Parmer | Item: 5014257
Ensure secure and leak-free sealing of sanitary ports
₹11,300.00INR / Each
₹11,300.00INR / Each
Product Description
Key Features
  • Cap off fluid system while maintaining cleanliness
  • Ideal for food and beverage, pharmaceutical, and biotechnology applications
  • ASME-BPE, USP Class VI, FDA 21 CFR 177.1520, REACH, and RoHS compliant
  • Animal-derived ingredient free (ADIF) and animal-derived component free (ADCF)
  • Polypropylene construction
  • 3" sanitary connection
More About this Item
Use sanitary end caps to seal inactive process line, preventing contamination and leakage. Sanitary end caps are often used in food and beverage, pharmaceutical, and biotechnology applications where maintaining cleanliness is crucial.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Sterilization MethodGamma irradiation and autoclave
  • Connection TypeEnd cap
  • Sanitary Clamp Size (Port 1) (InchShort)3
  • Regulatory ApprovalsRoHS;FDA;USP;ADCF
  • DescriptionSanitary End Cap, Polypropylene, 3"