Cole-Parmer SamplePrep 9001 Replacement Bowl for CG-900 Cryo-Blade Cryogenic Grinder, Stainless Steel, 1kg Sample Capacity
Representative image only.
Cole-Parmer SamplePrep 9001 Replacement Bowl for CG-900 Cryo-Blade Cryogenic Grinder, Stainless Steel, 1kg Sample Capacity
Cole-Parmer SamplePrep | Mfr: 9001| Item: 6123448
Optimize performance of your Cryo-Blade™ CG-900 Cryogenic Grinder
₹23,300.00INR / Each
₹23,300.00INR / Each
Product Description
Key Features
  • Removable 5 L inner bowl accommodates samples up to 1 kg
  • Allows samples to be stored in the freezer.
More About this Item
Enables rapid processing of samples from bulk to analytical preparation. Excellent for cryogenic grinding of fruits, vegetables, seeds, stalks, roots, fungi, and flowers.
Specifications
  • Compatible WithCG-900 Cryo-Blade™ Cryogenic Mill
  • Description9001 Cryogenic Processor Inner Bowl, 5 L 