Cole-Parmer SamplePrep 8507R Grinding Container Rack, Holds One or Three Grinding Containers
Representative image only.
Cole-Parmer SamplePrep 8507R Grinding Container Rack, Holds One or Three Grinding Containers
Cole-Parmer SamplePrep | Mfr: 8507R| Item: 0457824
Optimize performance of your Shatterbox® Verastile Ring and Puck Mill
₹132,000.00INR / Each
₹132,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer SamplePrep 8507R Grinding Container Rack, Holds One or Three Grinding Containers
Cole-Parmer SamplePrep 8507R Grinding Container Rack, Holds One or Three Grinding Containers
0457824
₹132,000.00INR / Each
+ + =
Total Price 798000.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Choose from a wide range of accessories and replacement parts to ensure safe operation
  • Required for grinding containers 04576-79 and 04576-80
  • Rack lid is matched to ShatterBox® mill clamp and holds grinding containers down during operation
More About this Item
Suitable for use with the Cole-Parmer® SM-200 ShatterBox® Ring and Puck Mills, 04578-25 and 04578-26. Mills sold separately.
Rack base plate has handles to lift rack into or out of Shatterbox® mill, and four locating pins for positioning grinding containers. When using with three grinding containers, all three should be the same kind 04576-79 or 04576-80.
Specifications
  • Compatible WithShatterBox® Ring and Puck Mill
  • DescriptionGrinding Container Rack, Holds One or Three Grinding Containers