Cole-Parmer SamplePrep 8016 Adapter for 35 mL Polypropylene Grinding Vial 04576-29; 1/EA
Representative image only.
Cole-Parmer SamplePrep 8016 Adapter for 35 mL Polypropylene Grinding Vial 04576-29; 1/EA
Cole-Parmer SamplePrep | Mfr: 8016| Item: 0457670
Use adapters to run multiple samples or to hold vial securely in place during grinding
₹14,500.00INR / Each
₹14,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer SamplePrep 8016 Adapter for 35 mL Polypropylene Grinding Vial 04576-29; 1/EA
Cole-Parmer SamplePrep 8016 Adapter for 35 mL Polypropylene Grinding Vial 04576-29; 1/EA
0457670
₹14,500.00INR / Each
+ =
Total Price 31400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
  • Required when running PP grinding vial 04576-29 in high-energy Mixer/Mill® grinders
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinder with a wide range of adapters and clamps.
Specifications
  • Compatible WithPolypropylene grinding vial 04576-29
  • DescriptionAdapter for 35 mL Polypropylene Grinding Vial 04576-29; 1/EA