Cole-Parmer SamplePrep 6871E Replacement End Plugs for Large Grinding Vial Set 61043-11, Low-Chrome Steel; 2/PK
Representative image only.
Cole-Parmer SamplePrep 6871E Replacement End Plugs for Large Grinding Vial Set 61043-11, Low-Chrome Steel; 2/PK
Cole-Parmer SamplePrep | Mfr: 6871E| Item: 6103462
Keep spare items to ensure there is no down time
₹91,400.00INR / Pkg of 2
₹91,400.00INR / Pkg of 2
Buy It Together
Current Item
Cole-Parmer SamplePrep 6871E Replacement End Plugs for Large Grinding Vial Set 61043-11, Low-Chrome Steel; 2/PK
Cole-Parmer SamplePrep 6871E Replacement End Plugs for Large Grinding Vial Set 61043-11, Low-Chrome Steel; 2/PK
6103462
₹91,400.00INR / Pkg of 2
+ + =
Total Price 105300.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Made of Cr-free ASTM 06 steel to eliminate chromium contaminationduring grinding
  • Specifically designed for grinding electronic components for RoHS/WEEE compliance testing
  • Autoclavable and reusable
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.
Specifications
  • MaterialLow-chrome steel
  • Compatible WithLarge grinding vial set 61043-11
  • DescriptionReplacement End Plugs for Large Grinding Vial Set 61043-11, Low-Chrome Steel; 2/PK