Cole-Parmer Safety Spectacles; 12 pcs/box
Representative image only.
Cole-Parmer Safety Spectacles; 12 pcs/box
Cole-Parmer | Item: 1639225
Eye protection with style and comfort
₹1,600.00INR / Box of 12
₹1,600.00INR / Box of 12
Product Description
Key Features
  • Extremely lightweight with outstanding field of vision
  • Single-piece lens fits most faces
  • Molded-in nose bridge provides uncompromised comfort and fit
  • Clear polycarbonate antiscratch lens with antifog coating
  • 99.99% UV protection
  • Spectacles feature Class 1 high optical quality lenses
  • 3 year warranty
More About this Item
Certifications:
• CE certified
• EN166:2001 compliant
• ANSI Z87.1-2015 compliant
3 Year WarrantyConformite Europeenne
Specifications
  • Frame ColorWhite
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceEN166:2001; ANSI Z87.1:2015
  • DescriptionSafety Spectacles; 12 pcs/box