Cole-Parmer Round Support Plate, 6" Dia Plate with 1/2" Dia Rod, Aluminum
Representative image only.
Cole-Parmer Round Support Plate, 6" Dia Plate with 1/2" Dia Rod, Aluminum
Cole-Parmer | Item: 0802716
Provide support for your specific laboratory equipment
₹14,900.00INR / Each
₹14,900.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Use to support your labware items from lab fames or support stands. The round plate design features a raised rim to prevent your apparatus from falling, requires an additional connector in order to attach clamps to frames or other apparatus. The rectangular plates include an integral connector.
NOTE: To install round plate to lab frames and support stands, you will need to purchase separately a connector that accepts a 0.5'' (12.7 cm) dia rod
1 Year Warranty
Specifications
  • Clamp MaterialAluminum
  • Diameter (in)6
  • DescriptionRound Support Plate, 6" Dia Plate with 1/2" Dia Rod, Aluminum