Cole-Parmer Roller Base for Dewars 03773-02, -03, and -04
Representative image only.
Cole-Parmer Roller Base for Dewars 03773-02, -03, and -04
Cole-Parmer | Item: 0377313
Safely and easily move cryogenic storage dewars with roller bases
₹102,000.00INR / Each
₹102,000.00INR / Each
Product Description
Key Features
  • Five casters provide stability
  • Sturdy aluminum construction
  • Lockable casters
  • Use with dewars 03773-02, -03, and -04
  • 3 month warranty
More About this Item
The sturdy aluminum roller bases feature five lockable casters for easy mobility and stability.
3 Month Warranty
Specifications
  • MaterialAluminum
  • Compatible WithDewars 03773-02, -03, and -04
  • DescriptionRoller Base for Dewars 03773-02, -03, and -04
  • Warranty3 months