Cole-Parmer RNA Avian Influenza A Virus Subtype H5, with mastermix
Representative image only.
Cole-Parmer RNA Avian Influenza A Virus Subtype H5, with mastermix
Cole-Parmer | Item: 9394787
Quickly screen for viral pathogens to prevent the spreading of disease causing systemic viruses
₹249,000.00INR / Each
₹249,000.00INR / Each
Product Description
Includes
Onestep mastermix (RNA transcriptase), lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control RNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Prevent economic loss, especially in the dairy and agricultural industry, using these viral detection kits to accurately assess whether these disease-causing pathogens are present. Select a complete kit with mastermix or order a kit without mastermix and use your own.
Specifications
  • Test TypeRNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • DescriptionAvian Influenza A Virus Subtype H5, with mastermix