Cole-Parmer Reversible 7-Point Horizontal Temperature Label, -30C-0C/-22-32F; 10/Pk

Representative image only.

Cole-Parmer Reversible 7-Point Horizontal Temperature Label, -30C-0C/-22-32F; 10/Pk

Cole-Parmer | Item: 0903706

Each style covers a wide range of temperatures, for use in many different application

₹2,400.00INR / Pkg of 10

₹2,400.00INR / Pkg of 10
Product Description

Key Features

  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty

More About this Item

These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.

1 Year Warranty
Specifications
  • Number of Temperature Indicators7
  • Max temperature (° F)32
  • Max temperature (° C)0
  • Temperature Points (° F)32 ,41, 50, 59, 68, 77, 86
  • Temperature Points (° C)0, 5, 10, 15, 20, 25, 30
  • Initial colorBlack
  • First color changeGreen (actual temperature); Brown (above actual); Blue (below actual)
  • Second color changeBlack
  • Height (in)1/2
  • Width (in)1 3/4
  • DescriptionReversible 7-Point Horizontal Temperature Label, -30C-0C/-22-32F; 10/Pk
  • Warranty1 year
Reviews
Q&A