Cole-Parmer Reversible 16-Point Vertical Temperature Label, -3-13C/26-56F; 10/Pk

Representative image only.

Cole-Parmer Reversible 16-Point Vertical Temperature Label, -3-13C/26-56F; 10/Pk

Cole-Parmer | Item: 0903719

Each style covers a wide range of temperatures, for use in many different application

₹2,600.00INR / Pkg of 10

₹2,600.00INR / Pkg of 10
Product Description

Key Features

  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty

More About this Item

These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.

1 Year Warranty
Specifications
  • Number of Temperature Indicators16
  • Max temperature (° F)56
  • Max temperature (° C)13
  • Temperature Points (° F)26 to 56
  • Temperature Points (° C)(-3), (-2), (-1), 0, 1, 2, 3, 4, 6, 7, 8, 9, 10, 11, 12, 13
  • Initial colorBlack
  • First color changeGreen (actual temperature); Brown (above actual); Blue (below actual)
  • Second color changeBlack
  • Height (in)5
  • Width (in)1/2
  • Depth (in)1/64
  • DescriptionReversible 16-Point Vertical Temperature Label, -3-13C/26-56F; 10/Pk
  • Warranty1 year
Reviews
Q&A